missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-serine benzyl ester, 95%

$75.26 - $259.08

Chemical Identifiers

CAS 59524-02-6
Molecular Formula C15H21NO5
Molecular Weight (g/mol) 295.335
MDL Number MFCD00076983
InChI Key BQADRZHPZVQGCW-LBPRGKRZSA-N
Synonym boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester, boc-ser-obzl
PubChem CID 7018792
IUPAC Name benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
SMILES CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5996503 View Documents Alfa Aesar™
H5996503
1g Each for $75.26
 
AAH5996506 View Documents Alfa Aesar™
H5996506
5g Each for $259.08
 
Description

Description

Applications
It is a pharmaceutical intermediate. It is involved in Mitsunobu Reactions.

Solubility
Slightly soluble in water.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

59524-02-6
295.335
BQADRZHPZVQGCW-LBPRGKRZSA-N
7018792
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
C15H21NO5
MFCD00076983
boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester, boc-ser-obzl
benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate

Specifications

70°C
59524-02-6
MFCD00076983
BQADRZHPZVQGCW-LBPRGKRZSA-N
benzyl (2S)-3-hydroxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoate
7018792
95%
1g
C15H21NO5
boc-ser-obzl, boc-l-serine benzyl ester, s-benzyl 2-tert-butoxycarbonyl amino-3-hydroxypropanoate, l-serine, n-1,1-dimethylethoxy carbonyl-, phenylmethyl ester, benzyl 2s-2-tert-butoxycarbonyl amino-3-hydroxypropanoate, ambotzbaa1202, pubchem12169, n-boc-serine benzyl ester, n-boc-l-serine benzyl ester, boc-ser-obzl
CC(C)(C)OC(=O)NC(CO)C(=O)OCC1=CC=CC=C1
295.335
295.33
N-Boc-L-serine benzyl ester