missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-prolinol, 98+%

Manufacturer:  Alfa Aesar™ L09885MD

 View more versions of this product

Catalog No. AAL09885MD



Specifications

Specifications

N-Boc-L-prolinol
59°C to 62°C
69610-40-8
MFCD00066232
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
CC(C)(C)OC(=O)N1CCCC1CO
201.266
201.27
−54° (c=5 in Methanol)
250mg
C10H19NO3
3542667
BFFLLBPMZCIGRM-QMMMGPOBSA-N
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
643448
≥98%

Chemical Identifiers

69610-40-8
201.266
BFFLLBPMZCIGRM-QMMMGPOBSA-N
643448
CC(C)(C)OC(=O)N1CCCC1CO
C10H19NO3
MFCD00066232
n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Safety and Handling

Safety and Handling

Recommended Storage : 2° to 8°C

Documents