Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-proline, 99%

$37.68 - $107.59

Chemical Identifiers

Chemical Name or Material N-Boc-L-proline
CAS 15761-39-4
Molecular Formula C10H17NO4
Formula Weight 215.25
MDL Number MFCD00037324
Beilstein 15828
Synonym Boc-Pro-OH; N-(tert-Butoxycarbonyl)-L-proline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1374406

  SDS
alfa aesar™
A1374406
5g Each for $37.68

AAA1374414

  SDS
alfa aesar™
A1374414
25g Each for $107.59
Description & Specifications

Specifications

Formula Weight 215.25
Optical Rotation −61° (c=1 in Acetic Acid)
MDL Number MFCD00037324
Assay 99%
Recommended Storage 2° to 8°C
Melting Point 132° to 136°C
Molecular Formula C10H17NO4
TSCA TSCA
CAS 15761-39-4
Beilstein 15828
Quantity 5g
Chemical Name or Material N-Boc-L-proline
Synonym Boc-Pro-OH; N-(tert-Butoxycarbonyl)-L-proline

Applications
N-Boc-L-proline is used as an intermediate in organic synthesis. It is also used to prepare daclatasvir, which inhibits the hepatitis C virus (HCV) non-structural 5A (NS5A) protein. Further, it is used as a drug in the treatment of hepatitis C virus (HCV).

Solubility
Soluble in acetic acid.

Notes
Moisture and light sensitive. Incompatible with strong oxidizing agents.

Safety and Handling

2° to 8°C