Learn More
N-Boc-L-prolinal, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H2649503
Description
Building block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBuilding block for novel nicotinic acetylcholine receptor ligands with cognition-enhancing properties. Used in the synthesis of chiral β-amino sulfides and β-amino thiols. Used to synthesize anticoagulants.
Solubility
Slightly soluble water. Soluble in chloroform.
Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, air, moisture.
Specifications
N-Boc-L-prolinal | |
1.06 | |
>110°C (230°F) | |
1.4589 | |
1 g | |
n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal | |
YDBPZCVWPFMBDH-QMMMGPOBSA-N | |
CC(C)(C)OC(=O)N1CCC[C@H]1C=O | |
199.25 | |
199.25 |
69610-41-9 | |
228°C | |
C10H17NO3 | |
MFCD00274186,MFCD00801206 | |
Air sensitive | |
Slightly soluble water. Soluble in chloroform. | |
−95° (c=1 in Chloroform) | |
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate | |
2734674 | |
96% |
Chemical Identifiers
69610-41-9 | |
199.25 | |
YDBPZCVWPFMBDH-QMMMGPOBSA-N | |
2734674 | |
CC(C)(C)OC(=O)N1CCC[C@H]1C=O |
C10H17NO3 | |
MFCD00274186,MFCD00801206 | |
n-boc-l-prolinal, boc-l-prolinal, s-1-boc-2-formylpyrrolidine, s-tert-butyl 2-formylpyrrolidine-1-carboxylate, tert-butyl 2s-2-formylpyrrolidine-1-carboxylate, boc-prolinal, n-tert-butoxycarbonyl-l-prolinal, s-n-tert-butoxycarbonyl-2-formylpyrrolidine, 1-pyrrolidinecarboxylic acid, 2-formyl-, 1,1-dimethylethyl ester, 2s, n-boc-prolinal | |
tert-butyl (2S)-2-formylpyrrolidine-1-carboxylate |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only