Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-phenylglycine, 99%

$59.41 - $195.49

Chemical Identifiers

Chemical Name or Material N-Boc-L-phenylglycine
CAS 2900-27-8
Molecular Formula C13H17NO4
Formula Weight 251.28
MDL Number MFCD00065588
Beilstein 3592362
Synonym Boc-Phg-OH; N-(tert-Butoxycarbonyl)-L-phenylglycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1854103

  SDS
alfa aesar™
L1854103
1g Each for $59.41

AAL1854106

  SDS
alfa aesar™
L1854106
5g Each for $195.49
Description & Specifications

Specifications

Formula Weight 251.28
Recommended Storage 2° to 8°C
Chemical Name or Material N-Boc-L-phenylglycine
Assay 99%
Molecular Formula C13H17NO4
MDL Number MFCD00065588
Melting Point 88° to 90°C
CAS 2900-27-8
Beilstein 3592362
Quantity 1g
Synonym Boc-Phg-OH; N-(tert-Butoxycarbonyl)-L-phenylglycine
Optical Rotation +141° (c=1 in Ethanol)
Safety and Handling

2° to 8°C