Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-phenylalanine 4-nitrophenyl ester, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-L-phenylalanine 4-nitrophenyl ester
CAS 7535-56-0
Formula Weight 386.4
MDL Number MFCD00042811
Beilstein 1895465
Synonym Boc-Phe-ONP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6676914

 
alfa aesar™
H6676914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6676922

 
alfa aesar™
H6676922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.4
Melting Point 127° to 128°C
Chemical Name or Material N-Boc-L-phenylalanine 4-nitrophenyl ester
Physical Form Crystalline Powder
Assay 97%
MDL Number MFCD00042811
Synonym Boc-Phe-ONP
Storage Note 1 -30° to -10°C
CAS 7535-56-0
Beilstein 1895465
Color White
Quantity 25g
Sensitivity Moisture Sensitive
Safety and Handling

-30° to -10°C