Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-phenylalaninal, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-L-phenylalaninal
CAS 72155-45-4
Formula Weight 249.31
MDL Number MFCD00143854
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6653003

 
alfa aesar™
H6653003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66530MD

 
alfa aesar™
H66530MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 249.31
Melting Point 85° to 89°C
Chemical Name or Material N-Boc-L-phenylalaninal
Assay 97%
MDL Number MFCD00143854
CAS 72155-45-4
Color White to pale brown
Physical Form Solid
Quantity 1g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C