Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-norvaline, 98+%

$169.97 - $169.97

Chemical Identifiers

Chemical Name or Material N-Boc-L-norvaline
CAS 53308-95-5
Molecular Formula C10H19NO4
Formula Weight 217.28g/mol
MDL Number MFCD00037268
Beilstein 3607582
Synonym Boc-Nva-OH; N-(tert-Butoxycarbonyl)-L-norvaline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0861503

 
alfa aesar™
L08615-03
1g Each for $169.97
Description & Specifications

Specifications

Quantity 1g
Melting Point 43° to 47°C
Quantity 1g
Optical Rotation −14° (c=1 in Methanol)
Chemical Name or Material N-Boc-L-norvaline
CAS 53308-95-5
Assay ≥98%
Molecular Formula C10H19NO4
Formula Weight 217.28g/mol
MDL Number MFCD00037268
Beilstein 3607582
Synonym Boc-Nva-OH; N-(tert-Butoxycarbonyl)-L-norvaline
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C