Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-norleucine, 98% (dry wt.), may cont. up to ca 10% solvent

$134.24 - $134.24

Chemical Identifiers

Chemical Name or Material N-Boc-L-norleucine
CAS 6404-28-0
Molecular Formula C11H21NO4
Formula Weight 231.29
MDL Number MFCD00037270
Beilstein 3608376
Synonym Boc-Nle-OH; N-(tert-Butoxycarbonyl)-L-norleucine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0853803

 
alfa aesar™
L0853803
1g Each for $134.24
Description & Specifications

Specifications

Formula Weight 231.29
Chemical Name or Material N-Boc-L-norleucine
Assay 98%
Molecular Formula C11H21NO4
MDL Number MFCD00037270
CAS 6404-28-0
Synonym Boc-Nle-OH; N-(tert-Butoxycarbonyl)-L-norleucine
Assay Percent Notes (dry wt.), may cont. up to ≈10% solvent
Beilstein 3608376
Quantity 1g
Optical Rotation −12.5° (c=1 in Acetic Acid)
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C