missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-methionine sulfoxide, 98%

Manufacturer:  Alfa Aesar™ H6305322

 View more versions of this product

Catalog No. AAH6305322



Specifications

Specifications

N-Boc-L-methionine sulfoxide
C10H19NO5S
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
CC(C)(C)OC(=O)NC(CCS(=O)C)C(=O)O
265.324
265.33
98%
34805-21-5
MFCD00037172
FVSDTYGQCVACMH-ISJKBYAMSA-N
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
7408090
Solid
100g

Chemical Identifiers

34805-21-5
265.324
FVSDTYGQCVACMH-ISJKBYAMSA-N
7408090
CC(C)(C)OC(=O)NC(CCS(=O)C)C(=O)O
C10H19NO5S
MFCD00037172
boc-met o-oh, boc-l-methionine sulfoxide, 2s-2-tert-butoxycarbonyl amino-4-methanesulfinylbutanoic acid, boc-met o, pubchem12247, n-alpha-t-butoxycarbonyl-l-methioninesulfoxid, n-alpha-tert-butyloxycarbonyl-l-methioninesulfoxide, 2s-2-tert-butoxycarbonylamino-4-methylsulfinyl-butanoic acid, 2s-2-tert-butoxy carbonyl amino-4-methanesulfinylbutanoic acid, 2s-2-2-methylpropan-2-yl oxycarbonylamino-4-methylsulfinylbutanoic acid
(2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-methylsulfinylbutanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

Documents