Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-methionine, 98+%

$26.36 - $46.26

Chemical Identifiers

Chemical Name or Material N-Boc-L-methionine
CAS 2488-15-5
Molecular Formula C10H19NO4S
Formula Weight 249.32
MDL Number MFCD00065586
Beilstein 1727869
Synonym Boc-Met-OH; N-(tert-Butoxycarbonyl)-L-methionine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0836603

  SDS
alfa aesar™
L0836603
1g Each for $26.36

AAL0836606

  SDS
alfa aesar™
L0836606
5g Each for $46.26
Description & Specifications

Specifications

Formula Weight 249.32
Chemical Name or Material N-Boc-L-methionine
MDL Number MFCD00065586
Molecular Formula C10H19NO4S
CAS 2488-15-5
Assay ≥98%
Synonym Boc-Met-OH; N-(tert-Butoxycarbonyl)-L-methionine
Recommended Storage 2° to 8°C
Optical Rotation −22° (c=1 in Methanol)
Odor Odorless
Quantity 1g
Beilstein 1727869
Melting Point 48° to 51°C
Safety and Handling

2° to 8°C