Please login to your online account to display your discounted pricing

N-BOC-L-Leucinol, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC376200050

 View more versions of this product

Catalog No. AC376200050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Viscous Liquid
Formula Weight 217.31
Quantity 5g
Color Clear colorless to pale yellow
Density 0.9830g/mL
Specific Rotation Condition - 27.00 (20.00°C C=2,methanol)
Chemical Name or Material N-BOC-L-Leucinol
Assay Percent Range 97%
CAS 82010-31-9
CAS Min % 96.0
CAS Max % 100.0
Specific Rotation -25° to -29° (20°C, 589nm) (C=2, methanol)
Infrared Spectrum Authentic
Molecular Formula C11H23NO3
MDL Number MFCD00076950
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)

Specifications

Chemical Name or Material N-BOC-L-Leucinol
CAS 82010-31-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H23NO3
Formula Weight 217.31
MDL Number MFCD00076950