Please login to your online account to display your discounted pricing

N-BOC-L-Leucinol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-BOC-L-Leucinol
CAS 82010-31-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H23NO3
Formula Weight 217.31
MDL Number MFCD00076950
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC376200050

  SDS
thermo fisher scientific
AC376200050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Viscous Liquid
Formula Weight 217.31
Quantity 5g
Color Clear colorless to pale yellow
Specific Rotation Condition - 27.00 (20.00°C C=2,methanol)
Density 0.9830g/mL
Assay Percent Range 97%
CAS 82010-31-9
Chemical Name or Material N-BOC-L-Leucinol
CAS Max % 100.0
CAS Min % 96.0
Specific Rotation -25° to -29° (20°C, 589nm) (C=2, methanol)
Packaging Glass bottle
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
Infrared Spectrum Authentic
Molecular Formula C11H23NO3
MDL Number MFCD00076950