Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-leucine methyl ester, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-L-leucine methyl ester
CAS 63096-02-6
Formula Weight 245.32
MDL Number MFCD00076952
Synonym Boc-Leu-OMe
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6155106

 
alfa aesar™
H6155106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6155114

 
alfa aesar™
H6155114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 245.32
Chemical Name or Material N-Boc-L-leucine methyl ester
Physical Form Liquid
Assay 97%
MDL Number MFCD00076952
CAS 63096-02-6
Synonym Boc-Leu-OMe
Refractive Index 1.4400
Density 0.991g/mL
Boiling Point 205°C
Quantity 5g