Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-isoleucine N-succinimidyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-isoleucine N-succinimidyl ester
CAS 3392-08-3
Formula Weight 328.37
MDL Number MFCD00022585
Beilstein 1552969
Synonym Boc-Ile-OSu; N-Boc-L-isoleucine N-hydroxysuccinimide ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6648006

  SDS
alfa aesar™
H6648006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6648014

  SDS
alfa aesar™
H6648014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6648022

  SDS
alfa aesar™
H6648022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 328.37
Melting Point 103° to 108°C
Chemical Name or Material N-Boc-L-isoleucine N-succinimidyl ester
Physical Form Powder
Assay 95%
CAS 3392-08-3
Synonym Boc-Ile-OSu; N-Boc-L-isoleucine N-hydroxysuccinimide ester
Quantity 5g
MDL Number MFCD00022585
Beilstein 1552969
Storage Note 1 -30° to -10°C
Color White
Safety and Handling

-30° to -10°C