missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nα-BOC-L-Histidine, 99+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC275590250

 View more versions of this product

Catalog No. AC275590250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Nα-BOC-L-Histidine
17791-52-5
100.0
C11H17N3O4
boc-his-oh, boc-l-his-oh, boc-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl, n-boc-l-histidine, n-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, tert-butyloxycarbonyl-l-histidine
AYMLQYFMYHISQO-QMMMGPOBSA-N
(2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
87308
+24.5° to +26.5° (20°C, 589nm) (c=1, CH3OH)
≥99.0%
White
99+%
99.0
99+%
Authentic
Glass bottle
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O
255.274
255.27
Powder, Crystals and/or Chunks
+25.5° (25°C c=1,methanol)
25g

Chemical Identifiers

17791-52-5
255.274
boc-his-oh, boc-l-his-oh, boc-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl, n-boc-l-histidine, n-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, tert-butyloxycarbonyl-l-histidine
(2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
C11H17N3O4
AYMLQYFMYHISQO-QMMMGPOBSA-N
87308
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O
Safety and Handling

Safety and Handling

EINECSNumber : 241-768-9

SDS Documents