missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nα-BOC-L-Histidine, 99+%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC275590010
Specifications
Nα-BOC-L-Histidine | |
17791-52-5 | |
100.0 | |
C11H17N3O4 | |
boc-his-oh, boc-l-his-oh, boc-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl, n-boc-l-histidine, n-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, tert-butyloxycarbonyl-l-histidine | |
AYMLQYFMYHISQO-QMMMGPOBSA-N | |
(2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid | |
87308 | |
+24.5° to +26.5° (20°C, 589nm) (c=1, CH3OH) | |
≥99.0% | |
White |
99+% | |
99.0 | |
99+% | |
Authentic | |
Glass bottle | |
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O | |
255.274 | |
255.27 | |
Powder, Crystals and/or Chunks | |
+25.5° (25°C c=1,methanol) | |
1g |
Chemical Identifiers
17791-52-5 | |
255.274 | |
boc-his-oh, boc-l-his-oh, boc-l-histidine, l-histidine, n-1,1-dimethylethoxy carbonyl, n-boc-l-histidine, n-tert-butoxycarbonyl-l-histidine, nalpha-boc-l-histidine, nalpha-tert-butoxycarbonyl-l-histidine, tert-butyloxycarbonyl-l-histidine | |
(2S)-3-(1H-imidazol-5-yl)-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid |
C11H17N3O4 | |
AYMLQYFMYHISQO-QMMMGPOBSA-N | |
87308 | |
CC(C)(C)OC(=O)NC(CC1=CN=CN1)C(=O)O |
Safety and Handling
EINECSNumber : 241-768-9