Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-glutamic acid 5-tert-butyl ester, 99%

$295.32 - $295.32

Chemical Identifiers

Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester
CAS 13726-84-6
Molecular Formula C14H25NO6
Formula Weight 303.36
MDL Number MFCD00038257
Synonym Boc-Glu(OtBu)-OH; N-tert-Butoxycarbonyl-L-glutamic acid 4-tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2232206

  SDS
alfa aesar™
B2232206
5g Each for $295.32
Description & Specifications

Specifications

Formula Weight 303.36
Quantity 5g
Molecular Formula C14H25NO6
Chemical Name or Material N-Boc-L-glutamic acid 5-tert-butyl ester
Assay 99%
CAS 13726-84-6
Melting Point 110° to 112°C
MDL Number MFCD00038257
Synonym Boc-Glu(OtBu)-OH; N-tert-Butoxycarbonyl-L-glutamic acid 4-tert-butyl ester