missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-glutamic acid 5-methyl ester, 95%

Manufacturer:  Alfa Aesar™ H62496MD

 View more versions of this product

Catalog No. AAH62496MD



Specifications

Specifications

N-Boc-L-glutamic acid 5-methyl ester
C11H19NO6
2s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid
CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)O
261.274
261.27
95%
250mg
45214-91-3
MFCD00190794
OHYMUFVCRVPMEY-ZETCQYMHSA-N
(2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
7018756
Solid
White

Chemical Identifiers

45214-91-3
261.274
OHYMUFVCRVPMEY-ZETCQYMHSA-N
7018756
CC(C)(C)OC(=O)NC(CCC(=O)OC)C(=O)O
C11H19NO6
MFCD00190794
2s-2-tert-butoxycarbonyl amino-5-methoxy-5-oxopentanoic acid
(2S)-5-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

Documents