Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-glutamic acid 5-cyclohexyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-glutamic acid 5-cyclohexyl ester
CAS 73821-97-3
Formula Weight 329.39
MDL Number MFCD00065570
Beilstein 3595818
Synonym Boc-Glu(OcHex)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6640906

 
alfa aesar™
H6640906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6640914

 
alfa aesar™
H6640914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6640922

 
alfa aesar™
H6640922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 329.39
Chemical Name or Material N-Boc-L-glutamic acid 5-cyclohexyl ester
CAS 73821-97-3
Assay 95%
Beilstein 3595818
Synonym Boc-Glu(OcHex)-OH
Physical Form Solid
MDL Number MFCD00065570
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C