missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-L-beta-leucine, 95%, Thermo Scientific Chemicals
$173.90 - $647.68
Chemical Identifiers
CAS | 183990-64-9 |
---|---|
Molecular Formula | C11H21NO4 |
Molecular Weight (g/mol) | 231.29 |
MDL Number | MFCD01076232 |
InChI Key | LUXMZCJCTUATDM-MRVPVSSYSA-N |
Synonym | boc-l-beta-homovaline, a-hoval-oh, r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid, boc-l-beta-hval-oh, boc-l-beta-hval-oh, r-n-boc-3-amino-4-methylpentanoic acid, r-n-boc-3-amino-4-methylpentanoic acid, boc-beta-leu-oh, ambotzbaa6280, boc-, boc- |
PubChem CID | 2761559 |
IUPAC Name | (3R)-3-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid |
SMILES | CC(C)[C@@H](CC(O)=O)NC(=O)OC(C)(C)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH66336MD
|
Thermo Scientific Chemicals
H66336MD |
250 mg |
Each for $173.90
|
|
|||||
AAH6633603
|
Thermo Scientific Chemicals
H6633603 |
1 g |
Each for $647.68
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
183990-64-9 | |
231.29 | |
LUXMZCJCTUATDM-MRVPVSSYSA-N | |
2761559 | |
CC(C)[C@@H](CC(O)=O)NC(=O)OC(C)(C)C |
C11H21NO4 | |
MFCD01076232 | |
boc-l-beta-homovaline, a-hoval-oh, r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid, boc-l-beta-hval-oh, boc-l-beta-hval-oh, r-n-boc-3-amino-4-methylpentanoic acid, r-n-boc-3-amino-4-methylpentanoic acid, boc-beta-leu-oh, ambotzbaa6280, boc-, boc- | |
(3R)-3-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid |
Specifications
115°C to 120°C | |
C11H21NO4 | |
7568192 | |
LUXMZCJCTUATDM-MRVPVSSYSA-N | |
(3R)-3-{[(tert-butoxy)carbonyl]amino}-4-methylpentanoic acid | |
2761559 | |
95% | |
Solid |
183990-64-9 | |
MFCD01076232 | |
boc-l-beta-homovaline, a-hoval-oh, r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid, boc-l-beta-hval-oh, boc-l-beta-hval-oh, r-n-boc-3-amino-4-methylpentanoic acid, r-n-boc-3-amino-4-methylpentanoic acid, 3r-3-tert-butoxycarbonyl amino-4-methylpentanoic acid, ambotzbaa6280, boc-, boc- | |
CC(C)[C@@H](CC(O)=O)NC(=O)OC(C)(C)C | |
231.29 | |
231.29 | |
250 mg | |
N-Boc-L-beta-leucine |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only