Please login to your online account to display your discounted pricing

N-BOC-L-Aspartic acid, 99%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC303210050

 View more versions of this product

Catalog No. AC303210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥98.5%
Formula Weight 233.22
Quantity 5g
Color White to off-white
Water 0.5% Max. (K.F.)
Chemical Name or Material N-BOC-L-Aspartic acid
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula HO2CCH2CHNHCO2C(CH3)3!CO2H
CAS 13726-67-5
Specific Rotation -4.5° to -6.5° (20°C, 589nm) (c=1, MeOH)
Molecular Formula C9H15NO6
Synonym N-(tert-Butoxycarbonyl)-L-aspartic acid

Specifications

Chemical Name or Material N-BOC-L-Aspartic acid
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C9H15NO6
Linear Formula HO2CCH2CHNHCO2C(CH3)3!CO2H
Formula Weight 233.22
Synonym N-(tert-Butoxycarbonyl)-L-aspartic acid