Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt
CAS 1913-12-8
Formula Weight 470.64
MDL Number MFCD00038889
Synonym Boc-Asp(OtBu)-OH.DCHA
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6162106

 
alfa aesar™
H6162106
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6162114

 
alfa aesar™
H6162114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 470.64
Chemical Name or Material N-Boc-L-aspartic acid 4-tert-butyl ester dicyclohexylammonium salt
Physical Form Solid
Assay 95%
MDL Number MFCD00038889
CAS 1913-12-8
Synonym Boc-Asp(OtBu)-OH.DCHA
Quantity 5g
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C