Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester
CAS 50715-50-9
Formula Weight 386.4
MDL Number MFCD00063389
Beilstein 5361818
Synonym Boc-Asp(OTBU)-OSU; (S)-2-[(Boc)(tert-butyl)amino]-4-(2,5-dioxopyrrolidin-1-yloxy)-4-oxobutyric acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6216303

  SDS
alfa aesar™
H6216303
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6216306

  SDS
alfa aesar™
H6216306
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 386.4
Melting Point 98° to 102°C
Chemical Name or Material N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester
Physical Form Powder
Assay 95%
MDL Number MFCD00063389
CAS 50715-50-9
Beilstein 5361818
Synonym Boc-Asp(OTBU)-OSU; (S)-2-[(Boc)(tert-butyl)amino]-4-(2,5-dioxopyrrolidin-1-yloxy)-4-oxobutyric acid
Color White
Quantity 1g
Sensitivity Moisture Sensitive
Storage Note 1 -30° to -10°C
Safety and Handling

-30° to -10°C