missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester, 95%

Manufacturer:  Alfa Aesar™ H6216306

 View more versions of this product

Catalog No. AAH6216306



Specifications

Specifications

N-Boc-L-aspartic acid 4-tert-butyl 1-(N-succinimidyl) ester
C17H26N2O8
5361818
PIITZDSTZQZNQH-JTQLQIEISA-N
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioate
13178396
Powder
White
5g
50715-50-9
MFCD00063389
1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-tert-butoxycarbonyl amino butanedioate
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
386.401
386.4
95%
98°C to 102°C
Moisture sensitive

Chemical Identifiers

50715-50-9
386.401
PIITZDSTZQZNQH-JTQLQIEISA-N
13178396
CC(C)(C)OC(=O)CC(C(=O)ON1C(=O)CCC1=O)NC(=O)OC(C)(C)C
C17H26N2O8
MFCD00063389
1-tert-butyl 2,5-dioxopyrrolidin-1-yl 3s-3-tert-butoxycarbonyl amino butanedioate
4-O-tert-butyl 1-O-(2,5-dioxopyrrolidin-1-yl) (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanedioate
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : -30° to -10°C

Recommended Storage : Store at -20°C; Store under Nitrogen

Documents