Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone

A caspase inhibitor designed to facilitate cell permeability

Chemical Identifiers

Chemical Name or Material N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone
CAS 187389-53-3
Formula Weight 263.26
MDL Number MFCD03453073
Synonym Boc-D-FMK; Boc-Asp(OMe)-FMK
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ65812LB0

 
alfa aesar™
J65812LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 263.26
Chemical Name or Material N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone
Physical Form Powder
MDL Number MFCD03453073
CAS 187389-53-3
Synonym Boc-D-FMK; Boc-Asp(OMe)-FMK
Solubility Information Soluble in dry DMSO or DMF at approximately 20mM; Store stock solutions at -20°C
Quantity 5mg
Storage Note 1 -30° to -10°C
Color Off-white

Applications
N-Boc-L-aspartic acid 4-methyl ester fluoromethyl ketone is used as a novel Boc-D(OMe)-FMK inhibitor synthesized using proprietary technology and irreversibly binds to activated caspases to block apoptosis. It is useful for in vitro and in vivo applications.

Solubility
Soluble in dry DMSO or DMF at approximately 20mM. Store stock solutions at -20°C.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

-30° to -10°C