Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 4-cyclohexyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-aspartic acid 4-cyclohexyl ester
CAS 73821-95-1
Formula Weight 315.36
MDL Number MFCD00061996
Beilstein 3563575
Synonym Boc-Asp(OcHx)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6124006

  SDS
alfa aesar™
H6124006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6124014

  SDS
alfa aesar™
H6124014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6124022

  SDS
alfa aesar™
H6124022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 93° to 95°C
Formula Weight 315.36
Chemical Name or Material N-Boc-L-aspartic acid 4-cyclohexyl ester
CAS 73821-95-1
Assay 95%
Synonym Boc-Asp(OcHx)-OH
Color White
Physical Form Powder
Beilstein 3563575
Quantity 5g
MDL Number MFCD00061996