missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 4-cyclohexyl ester, 95%

Manufacturer:  Alfa Aesar™ H6124022

 View more versions of this product

Catalog No. AAH6124022



Specifications

Specifications

N-Boc-L-aspartic acid 4-cyclohexyl ester
C15H25NO6
3563575
NLPQIWFEEKQBBN-NSHDSACASA-N
(2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
7009416
Powder
White
100g
73821-95-1
MFCD00061996
2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O
315.366
315.36
95%
93°C to 95°C

Chemical Identifiers

73821-95-1
315.366
NLPQIWFEEKQBBN-NSHDSACASA-N
7009416
CC(C)(C)OC(=O)NC(CC(=O)OC1CCCCC1)C(=O)O
C15H25NO6
MFCD00061996
2s-2-tert-butoxycarbonyl amino-4-cyclohexyloxy-4-oxobutanoic acid
(2S)-4-cyclohexyloxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

Documents