missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-L-aspartic acid 1-methyl ester, 98%, Thermo Scientific Chemicals
$102.26 - $1,165.92
Chemical Identifiers
CAS | 98045-03-5 |
---|---|
Molecular Formula | C10H17NO6 |
Molecular Weight (g/mol) | 247.247 |
MDL Number | MFCD03094777 |
InChI Key | IWFIVTBTZUCTQH-LURJTMIESA-N |
Synonym | boc-asp-ome, 1-methyl n-boc-l-aspartate, n-boc-l-aspartic acid 1-methyl ester, 3s-3-tert-butoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-boc asp oh ome, 1-methyl n-tert-butoxycarbonyl-l-aspartate, n-tert-butoxycarbonylaspartic acid a-methyl ester, n-tert-butoxycarbonyl aspartic acid 1-methyl ester, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl est, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl ester |
PubChem CID | 11021202 |
IUPAC Name | (3S)-4-methoxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62590MD
|
Thermo Scientific Chemicals
H62590MD |
250 mg |
N/A
|
|
|||||
AAH6259003
|
Thermo Scientific Chemicals
H6259003 |
1 g |
N/A
|
|
|||||
AAH6259006
|
Thermo Scientific Chemicals
H6259006 |
5 g |
N/A
|
|
|||||
Description
N-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Boc-L-aspartic acid 1-methyl ester is used as an intermediate in the preparation of modified alfa-amino acids and 5-cis-alkylprolines.
Notes
Incompatible with oxidizing agents. Heat sensitive.
Chemical Identifiers
98045-03-5 | |
247.247 | |
IWFIVTBTZUCTQH-LURJTMIESA-N | |
11021202 | |
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OC |
C10H17NO6 | |
MFCD03094777 | |
boc-asp-ome, 1-methyl n-boc-l-aspartate, n-boc-l-aspartic acid 1-methyl ester, 3s-3-tert-butoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-boc asp oh ome, 1-methyl n-tert-butoxycarbonyl-l-aspartate, n-tert-butoxycarbonylaspartic acid a-methyl ester, n-tert-butoxycarbonyl aspartic acid 1-methyl ester, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl est, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl ester | |
(3S)-4-methoxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Specifications
98045-03-5 | |
MFCD03094777 | |
IWFIVTBTZUCTQH-LURJTMIESA-N | |
(3S)-4-methoxy-3-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
11021202 | |
98% | |
Solid |
C10H17NO6 | |
boc-asp-ome, 1-methyl n-boc-l-aspartate, n-boc-l-aspartic acid 1-methyl ester, 3s-3-tert-butoxy carbonyl amino-4-methoxy-4-oxobutanoic acid, n-boc asp oh ome, 1-methyl n-tert-butoxycarbonyl-l-aspartate, n-tert-butoxycarbonylaspartic acid a-methyl ester, n-tert-butoxycarbonyl aspartic acid 1-methyl ester, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl est, n-tert-butoxycarbonyl-l-aspartic acid 1-methyl ester | |
CC(C)(C)OC(=O)NC(CC(=O)O)C(=O)OC | |
247.247 | |
247.25 | |
250 mg | |
N-Boc-L-aspartic acid 1-methyl ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only