Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-aspartic acid 1-benzyl ester, 99%

$88.97 - $88.97

Chemical Identifiers

Chemical Name or Material N-Boc-L-aspartic acid 1-benzyl ester
CAS 30925-18-9
Molecular Formula C16H21NO6
Formula Weight 323.35
MDL Number MFCD00065563
Synonym Boc-Asp-OBzl; N-tert-Butoxycarbonyl-L-aspartic acid 1-benzyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2231406

  SDS
alfa aesar™
B2231406
5g Each for $88.97
Description & Specifications

Specifications

Formula Weight 323.35
Chemical Name or Material N-Boc-L-aspartic acid 1-benzyl ester
Molecular Formula C16H21NO6
Melting Point 95° to 97°C
MDL Number MFCD00065563
CAS 30925-18-9
Synonym Boc-Asp-OBzl; N-tert-Butoxycarbonyl-L-aspartic acid 1-benzyl ester
Assay 99%
Quantity 5g

Applications
N-Boc-L-aspartic acid 1-benzyl ester is used as a local anesthetic and also used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in dichloromethane

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.