missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-BOC-L-Asparagine, 99%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC275510050

 View more versions of this product

Catalog No. AC275510050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

N-α-BOC-L-Asparagine
Glass bottle
White
5g
98.5
99%
NH2COCH2CH(COOH)NHCOOC(CH3)3
FYYSQDHBALBGHX-YFKPBYRVSA-N
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
82035
232.24
99%
Authentic
− 7.90
170.0°C to 175.0°C
7536-55-2
100.0
C9H16N2O5
boc-asn, boc-asn-oh, boc-asparagine, boc-l-asparagine, n-tert-butoxycarbonyl asparagine, n-tert-butoxycarbonyl-l-asparagine, nalpha-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, tert-butyloxycarbonyl-l-asparagine
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
232.24
CHEBI:3146
Powder or Crystalline Powder

Chemical Identifiers

7536-55-2
232.24
boc-asn, boc-asn-oh, boc-asparagine, boc-l-asparagine, n-tert-butoxycarbonyl asparagine, n-tert-butoxycarbonyl-l-asparagine, nalpha-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, tert-butyloxycarbonyl-l-asparagine
CHEBI:3146
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O
C9H16N2O5
FYYSQDHBALBGHX-YFKPBYRVSA-N
82035
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

EINECSNumber : 231-405-2

SDS Documents