Please login to your online account to display your discounted pricing

N-α-BOC-L-Asparagine, 99%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material N-α-BOC-L-Asparagine
CAS 7536-55-2
CAS Min % 98.5
CAS Max % 100.0
Assay 98 to 102%
Assay Percent Range 99%
Molecular Formula C9H16N2O5
Linear Formula NH2COCH2CH(COOH)NHCOOC(CH3)3
Formula Weight 232.24
Synonym BOC-Asn-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC275510050

  SDS
thermo fisher scientific
AC275510050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC275510250

  SDS
thermo fisher scientific
AC275510250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Powder or Crystalline Powder
Formula Weight 232.24
Quantity 5g
Color White
Assay 98 to 102%
CAS Max % 100.0
Chemical Name or Material N-α-BOC-L-Asparagine
Assay Percent Range 99%
Molecular Formula C9H16N2O5
CAS 7536-55-2
CAS Min % 98.5
Infrared Spectrum Authentic
Linear Formula NH2COCH2CH(COOH)NHCOOC(CH3)3
Synonym BOC-Asn-OH
Specific Rotation -6.5° to -8.5° (20°C, 589nm) (c=1 in DMF)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.