missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-α-BOC-L-Asparagine, 99%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC275510250
Specifications
N-α-BOC-L-Asparagine | |
− 7.90 | |
170.0°C to 175.0°C | |
7536-55-2 | |
100.0 | |
C9H16N2O5 | |
boc-asn, boc-asn-oh, boc-asparagine, boc-l-asparagine, n-tert-butoxycarbonyl asparagine, n-tert-butoxycarbonyl-l-asparagine, nalpha-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, tert-butyloxycarbonyl-l-asparagine | |
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O | |
232.24 | |
CHEBI:3146 | |
Powder or Crystalline Powder |
Authentic | |
White | |
25g | |
98.5 | |
99% | |
NH2COCH2CH(COOH)NHCOOC(CH3)3 | |
FYYSQDHBALBGHX-YFKPBYRVSA-N | |
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid | |
82035 | |
232.24 | |
99% |
Chemical Identifiers
7536-55-2 | |
232.24 | |
boc-asn, boc-asn-oh, boc-asparagine, boc-l-asparagine, n-tert-butoxycarbonyl asparagine, n-tert-butoxycarbonyl-l-asparagine, nalpha-tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonyl-l-asparagine, tert-butoxycarbonylasparagine, tert-butyloxycarbonyl-l-asparagine | |
CHEBI:3146 | |
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)O |
C9H16N2O5 | |
FYYSQDHBALBGHX-YFKPBYRVSA-N | |
82035 | |
(2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoic acid |
Safety and Handling
GHS Signal Word: Warning
EINECSNumber : 231-405-2