missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-α-BOC-L-Asparagine 4-nitrophenyl ester, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC440850050

 View more versions of this product

Catalog No. AC440850050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

N-α-BOC-L-Asparagine 4-nitrophenyl ester
-36.4
White
5g
97.5
97.5% min. (HPLC)
4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, boc-asn-onp, boc-asn-onp hplc, boc-asn-ophno2, boc-l-asparagine 4-nitrophenyl ester, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, pubchem12095, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, threonin-tert.-butylether
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
353.33
CHEBI:3147
Powder
Authentic
1% Max.
136°C to 140°C
4587-33-1
100.0
C15H19N3O7
IAPXDJMULQXGDD-NSHDSACASA-N
(4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate
439663
353.33
98%

Chemical Identifiers

4587-33-1
353.33
4-nitrophenyl 2s-2-tert-butoxycarbonyl amino-3-carbamoylpropanoate, boc-asn-onp, boc-asn-onp hplc, boc-asn-ophno2, boc-l-asparagine 4-nitrophenyl ester, ch3 3coconhch cooh ch2cooh, n1-t-boc c1-p-nitrophenol asparagine, pubchem12095, s-4-nitrophenyl 4-amino-2-tert-butoxycarbonyl amino-4-oxobutanoate, threonin-tert.-butylether
CHEBI:3147
CC(C)(C)OC(=O)NC(CC(=O)N)C(=O)OC1=CC=C(C=C1)[N+](=O)[O-]
C15H19N3O7
IAPXDJMULQXGDD-NSHDSACASA-N
439663
(4-nitrophenyl) (2S)-4-amino-2-[(2-methylpropan-2-yl)oxycarbonylamino]-4-oxobutanoate
Documents