Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-alpha-phenylglycinol, 98%

$109.92 - $374.04

Chemical Identifiers

Chemical Name or Material N-Boc-L-alpha-phenylglycinol
CAS 117049-14-6
Molecular Formula C13H19NO3
Formula Weight 237.3
MDL Number MFCD00274206
Beilstein 4316842
Synonym (S)-(+)-2-(Boc-amino)-2-phenylethanol; Boc-Phg-ol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1935303

  SDS
alfa aesar™
L1935303
1g Each for $109.92

AAL1935306

  SDS
alfa aesar™
L1935306
5g Each for $374.04
Description & Specifications

Specifications

Formula Weight 237.3
Chemical Name or Material N-Boc-L-alpha-phenylglycinol
Assay 98%
Molecular Formula C13H19NO3
Melting Point 136° to 140°C
CAS 117049-14-6
Beilstein 4316842
Synonym (S)-(+)-2-(Boc-amino)-2-phenylethanol; Boc-Phg-ol
Recommended Storage 2° to 8°C
MDL Number MFCD00274206
Quantity 1g
Optical Rotation +39° (c=1 in Chloroform)

Applications
N-Boc-L-alpha-phenylglycinol is an important raw material and intermediate used in organic Synthesis, pharmaceuticals, agrochemicals and dye fields. Phenylglycinol-derived oxazolopiperidone lactams is used as starting materials for the enantioselective synthesis of piperidine-containing alkaloids.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents. It is sensitive to moisture. Store under dry inert gas.

Safety and Handling

2° to 8°C