missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-allo-threonine dicyclohexylammonium salt, 98%

Chemical Identifiers

CAS 1464025-91-9
Formula Weight 400.56
MDL Number MFCD00038976
Synonym Boc-Allo-Thr-OH.DCHA
Chemical Name or Material N-Boc-L-allo-threonine dicyclohexylammonium salt
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6653703 Alfa Aesar™
H6653703
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAH66537MD Alfa Aesar™
H66537MD
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

1464025-91-9
MFCD00038976
N-Boc-L-allo-threonine dicyclohexylammonium salt
400.56
Boc-Allo-Thr-OH.DCHA

Specifications

400.56
MFCD00038976
Solid
N-Boc-L-allo-threonine dicyclohexylammonium salt
1464025-91-9
Boc-Allo-Thr-OH.DCHA
1g
98%