Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-L-alanine 4-nitrophenyl ester, 95%

Chemical Identifiers

Chemical Name or Material N-Boc-L-alanine 4-nitrophenyl ester
CAS 2483-49-0
Formula Weight 310.31
MDL Number MFCD00038117
Beilstein 1892073
Synonym Boc-Ala-ONP
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6677906

 
alfa aesar™
H6677906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6677914

 
alfa aesar™
H6677914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6677922

 
alfa aesar™
H6677922
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 310.31
Chemical Name or Material N-Boc-L-alanine 4-nitrophenyl ester
Synonym Boc-Ala-ONP
Physical Form Powder
Assay 95%
MDL Number MFCD00038117
Beilstein 1892073
Storage Note 1 2° to 8°C
CAS 2483-49-0
Color Pale brown
Quantity 5g
Sensitivity Light Sensitive
Safety and Handling

2° to 8°C