missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-hexahydro-1H-azepin-4-one, 98%

$62.68 - $626.81

Chemical Identifiers

CAS 188975-88-4
Molecular Formula C11H19NO3
Molecular Weight (g/mol) 213.277
MDL Number MFCD03788435
InChI Key PMLBUVZPRKXMOX-UHFFFAOYSA-N
Synonym 1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
PubChem CID 1512679
IUPAC Name tert-butyl 4-oxoazepane-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC(=O)CC1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH55214MD View Documents Alfa Aesar™
H55214MD
250mg Each for $62.68
 
AAH5521403 View Documents Alfa Aesar™
H5521403
1g Each for $180.00
 
AAH5521406 View Documents Alfa Aesar™
H5521406
5g Each for $626.81
 
Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.

Solubility
Soluble in Chloroform, Dichloromethane, Ethanol and Ethyl Ether.

Notes
Air sensitive. Keep under 2-8°C. Store under inert gas.
Specifications

Chemical Identifiers

188975-88-4
213.277
PMLBUVZPRKXMOX-UHFFFAOYSA-N
1512679
CC(C)(C)OC(=O)N1CCCC(=O)CC1
C11H19NO3
MFCD03788435
1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
tert-butyl 4-oxoazepane-1-carboxylate

Specifications

1.467
250mg
C11H19NO3
1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
PMLBUVZPRKXMOX-UHFFFAOYSA-N
tert-butyl 4-oxoazepane-1-carboxylate
1512679
98%
Air sensitive
188975-88-4
MFCD03788435
Soluble in Chloroform,Dichloromethane,Ethanol and Ethyl Ether.
CC(C)(C)OC(=O)N1CCCC(=O)CC1
213.277
213.28
N-Boc-hexahydro-1H-azepin-4-one
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Keep cold; Store under Argon