missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-hexahydro-1H-azepin-4-one, 98%

Manufacturer:  Alfa Aesar™ H55214MD

 View more versions of this product

Catalog No. AAH55214MD



Description

Description

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs. It is a fine chemical intermediate.

Solubility
Soluble in Chloroform, Dichloromethane, Ethanol and Ethyl Ether.

Notes
Air sensitive. Keep under 2-8°C. Store under inert gas.
Specifications

Specifications

N-Boc-hexahydro-1H-azepin-4-one
188975-88-4
MFCD03788435
PMLBUVZPRKXMOX-UHFFFAOYSA-N
tert-butyl 4-oxoazepane-1-carboxylate
1512679
98%
250mg
C11H19NO3
1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
CC(C)(C)OC(=O)N1CCCC(=O)CC1
213.277
213.28

Chemical Identifiers

188975-88-4
213.277
PMLBUVZPRKXMOX-UHFFFAOYSA-N
1512679
CC(C)(C)OC(=O)N1CCCC(=O)CC1
C11H19NO3
MFCD03788435
1,1-dimethylethyl 4-oxohexahydro-1h-azepine-1-carboxylate
tert-butyl 4-oxoazepane-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Recommended Storage : 2° to 8°C

Documents