Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-glycylglycine, 97%

Chemical Identifiers

Chemical Name or Material N-Boc-glycylglycine
CAS 31972-52-8
Linear Formula (CH3)3COCONHCH2CONHCH2CO2H
Formula Weight 232.23
MDL Number MFCD00038558
Synonym Boc-Gly-Gly-OH; N-(2-N-Boc-aminoacetyl)glycine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6288906

  SDS
alfa aesar™
H6288906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6288914

  SDS
alfa aesar™
H6288914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 232.23
MDL Number MFCD00038558
Chemical Name or Material N-Boc-glycylglycine
Assay 97%
CAS 31972-52-8
Synonym Boc-Gly-Gly-OH; N-(2-N-Boc-aminoacetyl)glycine
Linear Formula (CH3)3COCONHCH2CONHCH2CO2H
Color White
Storage Note 1 -30° to -10°C
Quantity 5g
Physical Form Powder
Safety and Handling

-30° to -10°C