Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-glycyl-L-leucine, 98%

Chemical Identifiers

Chemical Name or Material N-Boc-glycyl-L-leucine
CAS 51871-42-2
Formula Weight 288.34
MDL Number MFCD00190801
Synonym Boc-Gly-Leu-OH; (S)-2-(2-Boc-amino)acetamido-4-methylpentanoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6256903

  SDS
alfa aesar™
H6256903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6256906

  SDS
alfa aesar™
H6256906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH62569MD

  SDS
alfa aesar™
H62569MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 288.34
Chemical Name or Material N-Boc-glycyl-L-leucine
Physical Form Solid
Assay 98%
MDL Number MFCD00190801
CAS 51871-42-2
Synonym Boc-Gly-Leu-OH; (S)-2-(2-Boc-amino)acetamido-4-methylpentanoic acid
Quantity 1g