missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-glycyl-L-leucine, 98%

Manufacturer:  Alfa Aesar™ H6256906

 View more versions of this product

Catalog No. AAH6256906



Specifications

Specifications

N-Boc-glycyl-L-leucine
C13H24N2O5
boc-gly-leu-oh, l-leucine, n-1,1-dimethylethoxy carbonyl glycyl, boc-gly-l-leu-oh, 2s-2-2-tert-butoxycarbonyl amino acetamido-4-methylpentanoic acid
CC(C)CC(C(=O)O)NC(=O)CNC(=O)OC(C)(C)C
288.344
288.34
98%
51871-42-2
MFCD00190801
RGNNFKYULPHSJI-VIFPVBQESA-N
(2S)-4-methyl-2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]pentanoic acid
11953699
Solid
5g

Chemical Identifiers

51871-42-2
288.344
RGNNFKYULPHSJI-VIFPVBQESA-N
11953699
CC(C)CC(C(=O)O)NC(=O)CNC(=O)OC(C)(C)C
C13H24N2O5
MFCD00190801
boc-gly-leu-oh, l-leucine, n-1,1-dimethylethoxy carbonyl glycyl, boc-gly-l-leu-oh, 2s-2-2-tert-butoxycarbonyl amino acetamido-4-methylpentanoic acid
(2S)-4-methyl-2-[[2-[(2-methylpropan-2-yl)oxycarbonylamino]acetyl]amino]pentanoic acid
Documents