missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

N-Boc-glycine N-succinimidyl ester, 98%, Thermo Scientific Chemicals

$172.05 - $172.05

Chemical Identifiers

CAS 3392-07-2
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6252806
View Documents Promotion Details
Thermo Scientific Chemicals
H6252806
5 g
Each for $172.05
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

3392-07-2

Specifications

3392-07-2
C15H24N2O6
5 g
Hygroscopic
FATJLEZSGFVHQA-CABZTGNLSA-N
(2,5-dioxopyrrolidin-1-yl) (2R)-3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoate
131632401
98%
165°C to 167°C
MFCD00022585 MFCD00022585
1549242
boc-ile-osu, boc-l-isoleucine hydroxysuccinimide ester, boc-l-isoleucine n-hydroxysuccinimide ester, 2,5-dioxopyrrolidin-1-yl 2s,3s-2-tert-butoxycarbonyl amino-3-methylpentanoate, boc-lle-osu, boc-ile-osu hplc, n-alpha-t-butyloxycarbonyl-l-isoleucine succinimidyl ester, 1-n-1,1-dimethylethoxy carbonyl-l-isoleucyl oxy-2,5-pyrrolidinedione, 2,5-dioxopyrrolidin-1-yl 2s,3s-3-methyl-2-2-methylpropan-2-yl oxycarbonylamino pentanoate
CC[C@H](C)[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O
328.37
272.25
N-Boc-glycine N-succinimidyl ester
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 222-230-2

TSCA : No

Recommended Storage : Store at -20°C; Store under Argon

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only