Please login to your online account to display your discounted pricing

N-BOC-Glycine methyleester, 97%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC399380050

 View more versions of this product

Catalog No. AC399380050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 189.21
Quantity 5mL
Density 1.28g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material N-BOC-Glycine methyleester
Assay Percent Range 97%
Molecular Formula C8H15NO4
MDL Number MFCD00038267
CAS 31954-27-5
Packaging Glass bottle

Specifications

Chemical Name or Material N-BOC-Glycine methyleester
CAS 31954-27-5
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C8H15NO4
Formula Weight 189.21
MDL Number MFCD00038267