missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-DL-valine, 98%

$66.54 - $270.01

Chemical Identifiers

CAS 54895-12-4
Molecular Formula C10H19NO4
Molecular Weight (g/mol) 217.265
MDL Number MFCD00069973
InChI Key SZXBQTSZISFIAO-UHFFFAOYSA-N
Synonym 2-tert-butoxy carbonyl amino-3-methylbutanoic acid
PubChem CID 270655
IUPAC Name 3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)C(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6190806 View Documents Alfa Aesar™
H6190806
5g Each for $66.54
 
AAH6190814 View Documents Alfa Aesar™
H6190814
25g Each for $270.01
 
Specifications

Chemical Identifiers

54895-12-4
217.265
SZXBQTSZISFIAO-UHFFFAOYSA-N
270655
CC(C)C(C(=O)O)NC(=O)OC(C)(C)C
C10H19NO4
MFCD00069973
2-tert-butoxy carbonyl amino-3-methylbutanoic acid
3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

54895-12-4
MFCD00069973
2-tert-butoxy carbonyl amino-3-methylbutanoic acid
CC(C)C(C(=O)O)NC(=O)OC(C)(C)C
217.265
217.26
98%
5g
C10H19NO4
9196510
SZXBQTSZISFIAO-UHFFFAOYSA-N
3-methyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
270655
Solid
112°C to 114°C
N-Boc-DL-valine
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures