missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-DL-tert-leucine, 97%

$28.84 - $337.73

Chemical Identifiers

CAS 102185-35-3
Molecular Formula C11H21NO4
Molecular Weight (g/mol) 231.292
MDL Number MFCD00057782
InChI Key LRFZIPCTFBPFLX-UHFFFAOYSA-N
Synonym 2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-dl-tle-oh, n-boc-t-butylglycine, 2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid, 2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid, 2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-tbu-dl-gly-oh, boc-dl-tert-leucine, boc-dl-tbu gly-oh, n-boc-dl-tert-leucine
PubChem CID 5046859
IUPAC Name 3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
SMILES CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH66033MD View Documents Alfa Aesar™
H66033MD
250mg Each for $28.84
 
AAH6603303 View Documents Alfa Aesar™
H6603303
1g Each for $85.14
 
AAH6603306 View Documents Alfa Aesar™
H6603306
5g Each for $337.73
 
Specifications

Chemical Identifiers

102185-35-3
231.292
LRFZIPCTFBPFLX-UHFFFAOYSA-N
5046859
CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
C11H21NO4
MFCD00057782
2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-dl-tle-oh, n-boc-t-butylglycine, 2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid, 2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid, 2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-tbu-dl-gly-oh, boc-dl-tert-leucine, boc-dl-tbu gly-oh, n-boc-dl-tert-leucine
3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid

Specifications

102185-35-3
MFCD00057782
LRFZIPCTFBPFLX-UHFFFAOYSA-N
3,3-dimethyl-2-[(2-methylpropan-2-yl)oxycarbonylamino]butanoic acid
5046859
Solid
250mg
C11H21NO4
2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-dl-tle-oh, n-boc-t-butylglycine, 2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid, 2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid, 2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-tbu-dl-gly-oh, boc-dl-tert-leucine, boc-dl-tbu gly-oh, n-boc-dl-tert-leucine
CC(C)(C)C(C(=O)O)NC(=O)OC(C)(C)C
231.292
231.29
97%
N-Boc-DL-tert-leucine
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C