missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-DL-tert-leucine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H6603306
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
N-Boc-DL-tert-leucine | |
C11H21NO4 | |
2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-dl-tle-oh, n-boc-t-butylglycine, 2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid, 2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid, boc-tbu-dl-gly-oh, boc-dl-tert-leucine, boc-dl-tbu gly-oh, n-boc-dl-tert-leucine | |
CC(C)(C)OC(=O)NC(C(O)=O)C(C)(C)C | |
231.29 | |
231.29 | |
5 g |
169870-82-0 | |
MFCD00057782 | |
LRFZIPCTFBPFLX-UHFFFAOYNA-N | |
2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid | |
5046859 | |
97% | |
Solid |
Chemical Identifiers
169870-82-0 | |
231.29 | |
LRFZIPCTFBPFLX-UHFFFAOYNA-N | |
5046859 | |
CC(C)(C)OC(=O)NC(C(O)=O)C(C)(C)C |
C11H21NO4 | |
MFCD00057782 | |
2-tert-butoxycarbonyl amino-3,3-dimethylbutanoic acid, boc-dl-tle-oh, n-boc-t-butylglycine, 2-tert-butoxycarbonylamino-3,3-dimethyl-butyric acid, 2-tert-butoxycarbonylamino-3,3-dimethylbutanoic acid, boc-tbu-dl-gly-oh, boc-dl-tert-leucine, boc-dl-tbu gly-oh, n-boc-dl-tert-leucine | |
2-{[(tert-butoxy)carbonyl]amino}-3,3-dimethylbutanoic acid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold