missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-DL-phenylalanine, 95%

Manufacturer:  Alfa Aesar™ H6316403

 View more versions of this product

Catalog No. AAH6316403



Specifications

Specifications

N-Boc-DL-phenylalanine
C14H19NO4
boc-dl-phenylalanine, boc-dl-phe-oh, n-boc-dl-phenylalanine, n-tert-butoxycarbonyl phenylalanine, 2-tert-butoxy carbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonylamino-3-phenylpropanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, n-t-boc-phenylalanine
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)O
265.309
265.3
95%
4530-18-1
MFCD00558969
ZYJPUMXJBDHSIF-UHFFFAOYSA-N
2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
269698
Solid
1g

Chemical Identifiers

4530-18-1
265.309
ZYJPUMXJBDHSIF-UHFFFAOYSA-N
269698
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)O
C14H19NO4
MFCD00558969
boc-dl-phenylalanine, boc-dl-phe-oh, n-boc-dl-phenylalanine, n-tert-butoxycarbonyl phenylalanine, 2-tert-butoxy carbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonylamino-3-phenylpropanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, n-t-boc-phenylalanine
2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents