missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
N-Boc-DL-phenylalanine, 95%, Thermo Scientific Chemicals
$137.97 - $531.84
Chemical Identifiers
CAS | 4530-18-1 |
---|---|
Molecular Formula | C14H19NO4 |
Molecular Weight (g/mol) | 265.309 |
MDL Number | MFCD00558969 |
InChI Key | ZYJPUMXJBDHSIF-UHFFFAOYSA-N |
Synonym | boc-dl-phenylalanine, boc-dl-phe-oh, n-boc-dl-phenylalanine, n-tert-butoxycarbonyl phenylalanine, 2-tert-butoxy carbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonylamino-3-phenylpropanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoic acid, n-t-boc-phenylalanine |
PubChem CID | 269698 |
IUPAC Name | 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid |
SMILES | CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH6316406
|
Thermo Scientific Chemicals
H6316406 |
5 g |
Each for $137.97
|
|
|||||
AAH6316414
|
Thermo Scientific Chemicals
H6316414 |
25 g |
Each for $531.84
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
4530-18-1 | |
265.309 | |
ZYJPUMXJBDHSIF-UHFFFAOYSA-N | |
269698 | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)O |
C14H19NO4 | |
MFCD00558969 | |
boc-dl-phenylalanine, boc-dl-phe-oh, n-boc-dl-phenylalanine, n-tert-butoxycarbonyl phenylalanine, 2-tert-butoxy carbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonylamino-3-phenylpropanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoic acid, n-t-boc-phenylalanine | |
2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid |
Specifications
4530-18-1 | |
MFCD00558969 | |
ZYJPUMXJBDHSIF-UHFFFAOYSA-N | |
2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylpropanoic acid | |
269698 | |
95% | |
Solid |
C14H19NO4 | |
boc-dl-phenylalanine, boc-dl-phe-oh, n-boc-dl-phenylalanine, n-tert-butoxycarbonyl phenylalanine, 2-tert-butoxy carbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonyl amino-3-phenylpropanoic acid, 2-tert-butoxycarbonylamino-3-phenylpropanoic acid, 2-2-methylpropan-2-yl oxycarbonylamino-3-phenylpropanoic acid, n-t-boc-phenylalanine | |
CC(C)(C)OC(=O)NC(CC1=CC=CC=C1)C(=O)O | |
265.309 | |
265.3 | |
5 g | |
N-Boc-DL-phenylalanine |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only