Please login to your online account to display your discounted pricing

Alfa Aesar™ N-Boc-DL-nipecotic acid, 97%

$24.95 - $67.27

Chemical Identifiers

Chemical Name or Material N-Boc-DL-nipecotic acid
CAS 84358-12-3
Molecular Formula C11H19NO4
Formula Weight 229.28
MDL Number MFCD00673775
Beilstein 5539297
Synonym Boc-DL-Nip-OH; N-(tert-Butoxycarbonyl)nipecotic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1747803

  SDS
alfa aesar™
L1747803
1g Each for $24.95

AAL1747806

  SDS
alfa aesar™
L1747806
5g Each for $67.27
Description & Specifications

Specifications

Formula Weight 229.28
Chemical Name or Material N-Boc-DL-nipecotic acid
MDL Number MFCD00673775
CAS 84358-12-3
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 97%
Molecular Formula C11H19NO4
Melting Point 160° to 162°C
Beilstein 5539297
Quantity 1g
Synonym Boc-DL-Nip-OH; N-(tert-Butoxycarbonyl)nipecotic acid
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C